3'-Trifluoromethoxyacetophenone ≥97.0%

Supplier: TCI America

T1587-1G T1587-5G
TCT1587-1GEA 34.76 CAD
TCT1587-1G TCT1587-005G
3'-Trifluoromethoxyacetophenone ≥97.0%
3'-Trifluoromethoxyacetophenone

CAS Number: 170141-63-6
MDL Number: MFCD00042403
Molecular Formula: C9H7F3O2
Molecular Weight: 204.15
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 83
Specific Gravity (20/20): 1.29

Formula: C₉H₇F₃O₂
Density: 1.29 (20 °C)
Flash Pt: 83 °C
MDL Number: MFCD00042403
CAS Number: 170141-63-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4f236a7b32680e28229bf57d685ea3da7452ca75&vid=4f236a7b32680e28229bf57d685ea3da7452ca75&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR