1,2,3-Trichloro-5-nitrobenzene ≥96.0%

Supplier: TCI America

T1580-5G T1580-25G
TCT1580-005GEA 47.4 CAD
TCT1580-005G TCT1580-25G
1,2,3-Trichloro-5-nitrobenzene ≥96.0%
1,2,3-Trichloro-5-nitrobenzene

CAS Number: 20098-48-0
MDL Number: MFCD00061096
Molecular Formula: C6H2Cl3NO2
Molecular Weight: 226.44
Purity/Analysis Method: >96.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 71

Formula: C₆H₂Cl₃NO₂
Melting Pt: 71 °C
Density: 1.807 g/cm³ (20 °C)
MDL Number: MFCD00061096
CAS Number: 20098-48-0
EINECS: 243-511-6
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=75185241c2c60e8b8920b5e52aab8160a7e60636&vid=4746e81f19d7e6448d00e235b1de9c78b91173ea&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR