Tetrabutylammonium trifluoromethanesulfonate ≥97.0%

Supplier: TCI America

T1568-25G T1568-10G
TCT1568-25GEA 252.8 CAD
TCT1568-25G TCT1568-010G
Tetrabutylammonium trifluoromethanesulfonate ≥97.0%
Tetrabutylammonium trifluoromethanesulphonate

CAS Number: 35895-70-6
MDL Number: MFCD00042585
Molecular Formula: C17H36F3NO3S
Molecular Weight: 391.53
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Color: White
Melting point (°C): 113


Product Categories: Materials Chemistry, Secondary Battery Research Reagents
Other Product Names / Synonyms: Tetrabutylammonium Triflate, Trifluoromethanesulfonic Acid Tetrabutylammonium Salt

Formula: C₁₇H₃₆F₃NO₃S
MW: 391.54 g/mol
Melting Pt: 113 °C
MDL Number: MFCD00042585
CAS Number: 35895-70-6
EINECS: 252-783-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6985fc127e6f553cd4171229f1e3b4acd5ca4909&vid=6985fc127e6f553cd4171229f1e3b4acd5ca4909&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR