Sodium trifluoromethanesulfonate ≥98.0%

Supplier: TCI America

Synonyms: Trifluoromethanesulphonic acid sodium salt, Trifluoromethanesulfonic acid sodium salt

T1550-25G T1550-5G
TCT1550-025GEA 147.1 CAD
TCT1550-025G TCT1550-5G
Sodium trifluoromethanesulfonate ≥98.0%
Sodium trifluoromethanesulphonate

CAS Number: 2926-30-9
MDL Number: MFCD00061607
Molecular Formula: CHF3O3S
Molecular Weight: 172.05
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White


Product Categories: Synthetic Organic Chemistry, Homogeneous Catalysts, Homogeneous Catalysts
Other Product Names / Synonyms: Sodium Triflate, Trifluoromethanesulfonic Acid Sodium Salt

Formula: CF₃NaO₃S
MW: 172.06 g/mol
MDL Number: MFCD00061607
CAS Number: 2926-30-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=91384b45923b225b013f8e8f8809d9b3b58414e3&vid=91384b45923b225b013f8e8f8809d9b3b58414e3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR