3-(Trifluoromethyl)cinnamic acid ≥99.0%

Supplier: TCI America
Warning

Synonyms: 3-(3-(Trifluoromethyl)phenyl)acrylic acid

T1547-25G T1547-5G
TCT1547-025GEA 562.01 CAD
TCT1547-025G TCT1547-005G
3-(Trifluoromethyl)cinnamic acid ≥99.0%
3-(Trifluoromethyl)cinnamic acid

CAS Number: 779-89-5
MDL Number: MFCD00004393
Molecular Formula: C10H7F3O2
Molecular Weight: 216.16
Purity/Analysis Method: >99.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 135

Formula: C₁₀H₇F₃O₂
MW: 216.16 g/mol
Boiling Pt: 292
Melting Pt: 135 °C
MDL Number: MFCD00004393
CAS Number: 779-89-5
EINECS: 212-301-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=46b5361e380dc090cbc24f3c4332ff8aa33bee12&vid=46b5361e380dc090cbc24f3c4332ff8aa33bee12&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR