3'-(Trifluoromethyl)acetophenone ≥97.0%

Supplier: TCI America
Warning

T1546-25G T1546-5G
TCT1546-025GEA 137.46 CAD
TCT1546-025G TCT1546-005G
3'-(Trifluoromethyl)acetophenone ≥97.0%
3'-(Trifluoromethyl)acetophenone

CAS Number: 349-76-8
MDL Number: MFCD00000391
Molecular Formula: C9H7F3O
Molecular Weight: 188.15
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 199
Flash Point (°C): 83
Specific Gravity (20/20): 1.26


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 3-Acetylbenzotrifluoride

Formula: C₉H₇F₃O
Boiling Pt: 199 °C
Density: 1.26 (20 °C)
Flash Pt: 83 °C
Storage Temperature: Ambient
MDL Number: MFCD00000391
CAS Number: 349-76-8
EINECS: 206-490-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7b72df896ae0192a0316d5ff547fa23506023255&vid=7b72df896ae0192a0316d5ff547fa23506023255&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR