1,2,3-Trifluoro-4-nitrobenzene ≥98.0%
Supplier: TCI America
T1539-5G
T1539-25G
TCT1539-005GEA
50.56
CAD
TCT1539-005G
TCT1539-025G
1,2,3-Trifluoro-4-nitrobenzene ≥98.0%
1,2,3-Trifluoro-4-nitrobenzene
CAS Number: 771-69-7
MDL Number: MFCD00041546
Molecular Formula: C6H2F3NO2
Molecular Weight: 177.08
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Pale Yellow
Boiling point (°C): 92
Flash Point (°C): 93
Specific Gravity (20/20): 1.55
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 1,2,3-Trifluoro-4-nitrobenzene
Formula:
C₆H₂F₃NO₂ Boiling Pt: 92 °C Density: 1.55 (20 °C) Flash Pt: 93 °C |
MDL Number:
MFCD00041546 CAS Number: 771-69-7 EINECS: 212-238-4 UN: 2810 ADR: 6.1,III |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...