2-Trimethylsilyl-1,3-dithiane ≥96.0%

Supplier: TCI America

T1514-25G T1514-5G
TCT1514-025GEA 1448.86 CAD
TCT1514-025G TCT1514-005G
2-Trimethylsilyl-1,3-dithiane ≥96.0%
2-Trimethylsilyl-1,3-dithiane

CAS Number: 13411-42-2
MDL Number: MFCD00006655
Molecular Formula: C7H16S2Si
Molecular Weight: 192.41
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 77
Flash Point (°C): 96
Specific Gravity (20/20): 1.04


Product Categories: Synthetic Organic Chemistry, Sulfides, Carbon Homologation / Degradation Reaction, Silicon Compounds (for Synthesis)
Other Product Names / Synonyms: 1,3-Dithian-2-yltrimethylsilane

Formula: C₇H₁₆S₂Si
Boiling Pt: 77 °C
Density: 1.04 (20 °C)
Flash Pt: 96 °C
MDL Number: MFCD00006655
CAS Number: 13411-42-2
EINECS: 236-504-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4e2da42db22d06202aaaeea35caa0159d83bd446&vid=4e2da42db22d06202aaaeea35caa0159d83bd446&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR