Triethyl methanetricarboxylate ≥95.0%

Supplier: TCI America

T1481-5G T1481-25G
TCT1481-005GEA 86.9 CAD
TCT1481-005G TCT1481-25G
Triethyl methanetricarboxylate ≥95.0%
Triethyl methanetricarboxylate

CAS Number: 6279-86-3
MDL Number: MFCD00009150
Molecular Formula: C10H16O6
Molecular Weight: 232.23
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 253
Flash Point (°C): 113
Freezing point (°C): 25


Other Product Names / Synonyms: Carboxymalonic Acid Triethyl Ester, Methanetricarboxylic Acid Triethyl Ester, Triethyl Carboxymalonate

Formula: C₁₀H₁₆O₆
MW: 232.23 g/mol
Boiling Pt: 253 °C
Flash Pt: 113 °C
MDL Number: MFCD00009150
CAS Number: 6279-86-3
EINECS: 228-477-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=64efc3b0fde3dd163383fa8444d75d1cc23f715f&vid=64efc3b0fde3dd163383fa8444d75d1cc23f715f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR