2,4,6-Trimethylbenzoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

T1421-10G T1421-25G
TCT1421-010GEA 47.4 CAD
TCT1421-010G TCT1421-25G
2,4,6-Trimethylbenzoic acid ≥98.0% (by GC, titration analysis)
2,4,6-Trimethylbenzoic acid

CAS Number: 480-63-7
MDL Number: MFCD00002481
Molecular Formula: C10H12O2
Molecular Weight: 164.20
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 154


Other Product Names / Synonyms: Mesitylenecarboxylic Acid

Formula: C₁₀H₁₂O₂
Boiling Pt: 298 °C (1013 hPa)
Melting Pt: 154 °C
Storage Temperature: Ambient
MDL Number: MFCD00002481
CAS Number: 480-63-7
EINECS: 207-553-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ae26bc2edb09d17c7a92d0f20666ae24e98928a7&vid=e092cb655c32b99099c83a997116a788987f06cb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR