4-(Trifluoromethoxy)benzoic acid ≥97.0%

Supplier: TCI America
Warning

T1400-5G T1400-1G
TCT1400-005GEA 123.24 CAD
TCT1400-005G TCT1400-001G
4-(Trifluoromethoxy)benzoic acid ≥97.0%
4-(Trifluoromethoxy)benzoic acid

CAS Number: 330-12-1
MDL Number: MFCD00002541
Molecular Formula: C8H5F3O3
Molecular Weight: 206.12
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 154


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: alpha,alpha,alpha-Trifluoro-p-anisic Acid

Formula: C₈H₅F₃O₃
MW: 206.12 g/mol
Boiling Pt: 272
Melting Pt: 153.4
MDL Number: MFCD00002541
CAS Number: 330-12-1
EINECS: 206-352-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=23b2d4c4379c6e9c066dcedf321c2cf50a8252e4&vid=e8de06a925cb0808143e2738ec6640ca0dbb07fb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR