α,α,α-Trifluoro-3-tolualdehyde ≥95.0%

Supplier: TCI America
Warning

Synonyms: 3-(Trifluoromethyl)benzaldehyde, Cinacalcet Impurity 23

T1399-5G T1399-25G
TCT1399-005GEA 52.93 CAD
TCT1399-005G TCT1399-025G
α,α,α-Trifluoro-3-tolualdehyde ≥95.0%
α,α,α-Trifluoro-3-tolualdehyde

CAS Number: 454-89-7
MDL Number: MFCD00003373
Molecular Formula: C8H5F3O
Molecular Weight: 174.12
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 66
Flash Point (°C): 68
Specific Gravity (20/20): 1.31


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: alpha,alpha,alpha-Trifluoro-m-tolualdehyde

Formula: C₈H₅F₃O
MW: 174.12 g/mol
Boiling Pt: 66 °C
Density: 1.31 (20 °C)
Flash Pt: 68 °C
MDL Number: MFCD00003373
CAS Number: 454-89-7
EINECS: 207-228-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4837108d8a6741f576cc0acac8045961cd0f5aae&vid=4837108d8a6741f576cc0acac8045961cd0f5aae&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR