2,3,4,5-Tetrafluorobenzoic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

T1395-25G T1395-5G
TCT1395-025GEA 37.92 CAD
TCT1395-025G TCT1395-005G
2,3,4,5-Tetrafluorobenzoic acid ≥98.0% (by HPLC, titration analysis)
2,3,4,5-Tetrafluorobenzoic acid

CAS Number: 1201-31-6
MDL Number: MFCD00009613
Molecular Formula: C7H2F4O2
Molecular Weight: 194.08
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: White
Melting point (°C): 87

Formula: C₇H₂F₄O₂
Boiling Pt: 135…137 °C (23 mmHg)
Melting Pt: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD00009613
CAS Number: 1201-31-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6b02df97d257cd1f4adc576487864c34dccc7257&vid=eb7669acab72b6f75b5874f7f68fb1c5f79f4cbf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR