Tetraphenylphosphonium chloride ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
T1375-25G
T1375-5G
TCT1375-025GEA
257.75
CAD
TCT1375-025G
TCT1375-005G
Tetraphenylphosphonium chloride ≥98.0% (by HPLC, titration analysis)
Tetraphenylphosphonium chloride
CAS Number: 2001-45-8
MDL Number: MFCD00011916
Molecular Formula: C24H20ClP
Molecular Weight: 374.85
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: White
Formula:
C₂₄H₂₀ClP Melting Pt: 275…278 °C |
MDL Number:
MFCD00011916 CAS Number: 2001-45-8 EINECS: 217-890-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...