Tetrakis(triphenylphosphine)palladium(0) ≥97.0%
Supplier: TCI America
Synonyms:
Tetrakis(triphenylphosphine)palladium, Pd(PPh3)4
T1350-1G
T1350-25G
T1350-5G
TCT1350-001GEA
60.04
CAD
TCT1350-001G
TCT1350-25G
TCT1350-005G
Tetrakis(triphenylphosphine)palladium(0) ≥97.0%
Tetrakis(triphenylphosphine)palladium (0)
CAS Number: 14221-01-3
MDL Number: MFCD00010012
Molecular Formula: C72H60P4Pd
Molecular Weight: 1155.59
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Color: Pale Yellow
Melting point (°C): 105
Storage Temperature: >-20°C
Formula:
C₇₂H₆₀P₄Pd Melting Pt: 105 °C Density: 1 g/cm³ (20 °C) |
MDL Number:
MFCD00010012 CAS Number: 14221-01-3 EINECS: 238-086-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...