3-(Trifluoromethyl)benzyl alcohol ≥98.0%
Supplier: TCI America
T1342-10G
T1342-25G
TCT1342-010GEA
107.44
CAD
TCT1342-010G
TCT1342-25G
3-(Trifluoromethyl)benzyl alcohol ≥98.0%
3-(Trifluoromethyl)benzyl alcohol
CAS Number: 349-75-7
MDL Number: MFCD00004645
Molecular Formula: C8H7F3O
Molecular Weight: 176.14
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 68
Flash Point (°C): 84
Specific Gravity (20/20): 1.30
Formula:
C₈H₇F₃O Boiling Pt: 68 °C Density: 1.30 (20 °C) Flash Pt: 84 °C |
MDL Number:
MFCD00004645 CAS Number: 349-75-7 EINECS: 206-489-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...