Sodium trifluoroacetate ≥98.0%
Supplier: TCI America
Synonyms:
Sodium 2,2,2-trifluoroacetate
T1336-25G
T1336-500G
T1336-100G
TCT1336-025GEA
70.45
CAD
TCT1336-025G
TCT1336-500G
TCT1336-100G
Sodium trifluoroacetate ≥98.0%
Sodium trifluoroacetate
CAS Number: 2923-18-4
MDL Number: MFCD00013217
Molecular Formula: C2HF3O2
Molecular Weight: 136.00
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 207
Product Categories: Synthetic Organic Chemistry, Fluorinating Agents, Building Blocks, Fluorinated Bioactive Compounds, Difluoromethylating Agents
Other Product Names / Synonyms: Trifluoroacetic Acid Sodium Salt
Formula:
C₂F₃NaO₂ Melting Pt: 207 °C |
MDL Number:
MFCD00013217 CAS Number: 2923-18-4 EINECS: 220-879-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...