Silver trifluoromethanesulfonate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Silver trifluoromethanesulphonate, Silver triflate, Silver trifluoromethanesulfonate, Trifluoromethanesulphonic acid silver salt, Trifluoromethanesulfonic acid silver salt

T1331-10G T1331-25G T1331-1G
TCT1331-010GEA 215.72 CAD
TCT1331-010G TCT1331-25G TCT1331-1G
Silver trifluoromethanesulfonate ≥98.0% (by titrimetric analysis)
Silver trifluoromethanesulphonate

CAS Number: 2923-28-6
MDL Number: MFCD00013226
Molecular Formula: CHF3O3S
Molecular Weight: 256.93
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White


Product Categories: Synthetic Organic Chemistry, Homogeneous Catalysts, Homogeneous Catalysts, Glycosylation, Stable Lewis Acids in Aqueous Media, Additives ([2+2+2] Cycloaddition Reaction)
Other Product Names / Synonyms: Silver Triflate, Trifluoromethanesulfonic Acid Silver Salt

Formula: CAgF₃O₃S
MW: 256.94 g/mol
Melting Pt: 356 °C
MDL Number: MFCD00013226
CAS Number: 2923-28-6
EINECS: 220-882-2
UN: 3260
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d17f1a629c79ac4742c87b6433934db06d677af5&vid=d17f1a629c79ac4742c87b6433934db06d677af5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR