4-(Trifluoromethyl)benzyl alcohol ≥96.0%

Supplier: TCI America
Warning

Synonyms: (4-(Trifluoromethyl)phenyl)methanol, 4-(Hydroxymethyl)benzotrifluoride

T1312-5G T1312-25G
TCT1312-005GEA 63.2 CAD
TCT1312-005G TCT1312-025G
4-(Trifluoromethyl)benzyl alcohol ≥96.0%
4-(Trifluoromethyl)benzyl alcohol

CAS Number: 349-95-1
MDL Number: MFCD00004661
Molecular Formula: C8H7F3O
Molecular Weight: 176.14
Purity/Analysis Method: >96.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 80
Flash Point (°C): 100
Freezing point (°C): 24

Formula: C₈H₇F₃O
MW: 176.14 g/mol
Boiling Pt: 214
Melting Pt: 23
Density: 1.28 g/cm³ (20 °C)
Flash Pt: 100 °C
MDL Number: MFCD00004661
CAS Number: 349-95-1
EINECS: 206-494-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ec205407cef7b67df495fb61d391f9c24d49dc0a&vid=ec205407cef7b67df495fb61d391f9c24d49dc0a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR