Trifluoromethanesulfonamide ≥98.0% (by titrimetric analysis)

Supplier: TCI America

T1290-25G T1290-10G
TCT1290-25GEA 453.9 CAD
TCT1290-25G TCT1290-010G
Trifluoromethanesulfonamide ≥98.0% (by titrimetric analysis)
Trifluoromethanesulphonamide

CAS Number: 421-85-2
MDL Number: MFCD00068714
Molecular Formula: CH2F3NO2S
Molecular Weight: 149.09
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 120

Formula: CH₂F₃NO₂S
Melting Pt: 120 °C
MDL Number: MFCD00068714
CAS Number: 421-85-2
EINECS: 431-270-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1815d5c07618129b7966f9d01fd57541fad6daa8&vid=1815d5c07618129b7966f9d01fd57541fad6daa8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR