Methyl-m-toluate ≥98.0%

Supplier: TCI America

T1258-25ML T1258-500ML
TCT1258-025MLEA 55.3 CAD
TCT1258-025ML TCT1258-500ML
Methyl-m-toluate ≥98.0%
Methyl m-toluate

CAS Number: 99-36-5
MDL Number: MFCD00008436
Molecular Formula: C9H10O2
Molecular Weight: 150.18
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 115
Flash Point (°C): 96
Specific Gravity (20/20): 1.06


Other Product Names / Synonyms: 3-Methylbenzoic Acid Methyl Ester, m-Toluic Acid Methyl Ester

Formula: C₉H₁₀O₂
Boiling Pt: 115 °C
Density: 1.06 (20 °C)
Flash Pt: 96 °C
MDL Number: MFCD00008436
CAS Number: 99-36-5
EINECS: 202-753-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5f45e2650910b66ab9c0db5d6d946bd9a7e1feb0&vid=5f45e2650910b66ab9c0db5d6d946bd9a7e1feb0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR