Methyl-p-benzoquinone ≥98.0%

Supplier: TCI America
Warning

Synonyms: 2-Methylcyclohexa-2,5-diene-1,4-dione, Methylquinone, Methyl-1,4-benzoquinone

T1244-25G T1244-500G
TCT1244-025GEA 105.86 CAD
TCT1244-025G TCT1244-500G
Methyl-p-benzoquinone ≥98.0%
Methyl-p-benzoquinone

CAS Number: 553-97-9
MDL Number: MFCD00001603
Molecular Formula: C7H6O2
Molecular Weight: 122.12
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: Pale Yellow
Melting point (°C): 68


Product Categories: Synthetic Organic Chemistry, C-H Bond Activation Reaction
Other Product Names / Synonyms: Methyl-p-benzoquinone

Formula: C₇H₆O₂
MW: 122.12 g/mol
Boiling Pt: 187
Melting Pt: 68 °C
Density: 1.08 g/cm³ (75.5 °C)
MDL Number: MFCD00001603
CAS Number: 553-97-9
EINECS: 209-056-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e68b2e5bb4c5f7e9d47d2de7b3ad21145a95d5dc&vid=e68b2e5bb4c5f7e9d47d2de7b3ad21145a95d5dc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR