Ethyl-p-tolylacetate ≥98.0%

Supplier: TCI America

Synonyms: Ethyl p-Tolylacetate, (4-Methylphenyl)acetic acid ethyl ester, p-Tolylacetic acid ethyl ester

T1227-5G T1227-25G
TCT1227-005GEA 85.21 CAD
TCT1227-005G TCT1227-025G
Ethyl-p-tolylacetate ≥98.0%
Ethyl p-tolylacetate

CAS Number: 14062-19-2
MDL Number: MFCD00009181
Molecular Formula: C11H14O2
Molecular Weight: 178.23
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 240
Flash Point (°C): 109
Specific Gravity (20/20): 1.01


Other Product Names / Synonyms: Ethyl 4-Methylphenylacetate, p-Tolylacetic Acid Ethyl Ester, 4-Methylphenylacetic Acid Ethyl Ester

Formula: (C₆H₄(CH₃))CH₂COOCH₂CH₃
MW: 178.23 g/mol
Boiling Pt: 240 °C
Density: 1.01 (20 °C)
Flash Pt: 109 °C
MDL Number: MFCD00009181
CAS Number: 14062-19-2
EINECS: 237-904-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=48eb3d20a7c942a04c9643ae99c8c4fc889f342b&vid=48eb3d20a7c942a04c9643ae99c8c4fc889f342b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR