Ethyl-O-tolylacetate ≥97.0%

Supplier: TCI America

T1225-25G T1225-5G
TCT1225-025GEA 188.02 CAD
TCT1225-025G TCT1225-005G
Ethyl-O-tolylacetate ≥97.0%
Ethyl o-tolylacetate

CAS Number: 40291-39-2
MDL Number: MFCD00009179
Molecular Formula: C11H14O2
Molecular Weight: 178.23
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 77
Flash Point (°C): 107
Specific Gravity (20/20): 1.02


Other Product Names / Synonyms: Ethyl 2-Methylphenylacetate, o-Tolylacetic Acid Ethyl Ester, 2-Methylphenylacetic Acid Ethyl Ester

Formula: (C₆H₄CH₃)CH₂COOCH₂CH₃
Boiling Pt: 77 °C
Density: 1.02 (20 °C)
Flash Pt: 107 °C
MDL Number: MFCD00009179
CAS Number: 40291-39-2
EINECS: 254-871-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b17eb34170bee2e3841573a44bd8b8f9e373d46b&vid=b17eb34170bee2e3841573a44bd8b8f9e373d46b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR