2',3',4'-Trihydroxyacetophenone ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

T1198-5G T1198-25G
TCT1198-005GEA 58.46 CAD
TCT1198-005G TCT1198-025G
2',3',4'-Trihydroxyacetophenone ≥98.0% (by HPLC, titration analysis)
2',3',4'-Trihydroxyacetophenone

CAS Number: 528-21-2
MDL Number: MFCD00002193
Molecular Formula: C8H8O4
Molecular Weight: 168.15
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: White
Melting point (°C): 170


Other Product Names / Synonyms: Gallacetophenone

Formula: C₈H₈O₄
Melting Pt: 170 °C
MDL Number: MFCD00002193
CAS Number: 528-21-2
EINECS: 208-430-2
Merck Index: 12,04360
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b9acae19148f4302ebe5f810a4b71b06057d77c1&vid=b9acae19148f4302ebe5f810a4b71b06057d77c1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR