2,2':5',2''-Terthiophene ≥98.0%, purified by sublimation

Supplier: TCI America

T1196-1G T1196-5G
TCT1196-001GEA 113.76 CAD
TCT1196-001G TCT1196-005G
2,2':5',2''-Terthiophene ≥98.0%, purified by sublimation
2,2':5',2''-Terthiophene

CAS Number: 1081-34-1
MDL Number: MFCD00012167
Molecular Formula: C12H8S3
Molecular Weight: 248.38
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: Slightly Pale Yellow
Melting point (°C): 95


Product Categories: Materials Chemistry, Organic Transistor Materials, Conducting Polymer Research Reagents, Highly Purified Reagents
Other Product Names / Synonyms: 2,5-Di(2-thienyl)thiophene, alpha-Terthienyl

Formula: C₁₂H₈S₃
Melting Pt: 95 °C
MDL Number: MFCD00012167
CAS Number: 1081-34-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d1bd58844f56db339d569a1e303504467fbb4f2d&vid=13f6421bc58341d790aa794e214e695ce86f774d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR