D-(-)-Diethyl tartrate ≥97.0%
Supplier: TCI America
T1195-25G
T1195-250G
TCT1195-025GEA
36.34
CAD
TCT1195-025G
TCT1195-250G
D-(-)-Diethyl tartrate ≥97.0%
D(-)-Diethyl tartrate
CAS Number: 13811-71-7
MDL Number: MFCD00064451
Molecular Formula: C8H14O6
Molecular Weight: 206.19
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 130
Flash Point (°C): 93
Specific Gravity (20/20): 1.21
Specific rotation [a]20/D: -7.8 deg (neat)
Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: D-(-)-Tartaric Acid Diethyl Ester
Formula:
C₈H₁₄O₆ Boiling Pt: 130 °C Melting Pt: 17 °C Density: 1.21 (20 °C) Flash Pt: 93 °C |
MDL Number:
MFCD00064451 CAS Number: 13811-71-7 EINECS: 237-458-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...