2,4,5-Trimethoxybenzaldehyde ≥98.0%
Supplier: TCI America
T1172-5G
T1172-25G
TCT1172-005GEA
26.86
CAD
TCT1172-005G
TCT1172-025G
2,4,5-Trimethoxybenzaldehyde ≥98.0%
2,4,5-Trimethoxybenzaldehyde
CAS Number: 4460-86-0
MDL Number: MFCD00003312
Molecular Formula: C10H12O4
Molecular Weight: 196.20
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: Very Pale Yellow
Boiling point (°C): 140
Melting point (°C): 113
Formula:
C₁₀H₁₂O₄ MW: 196.2 g/mol Boiling Pt: 140 °C Melting Pt: 113 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003312 CAS Number: 4460-86-0 EINECS: 224-713-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...