1-Amino-2,2,2-trifluoroethane hydrochloride ≥98.0% (by total nitrogen and titration analysis)
Supplier: TCI America
T1170-25G
T1170-5G
TCT1170-025GEA
102.86
CAD
TCT1170-025G
TCT1170-005G
1-Amino-2,2,2-trifluoroethane hydrochloride ≥98.0% (by total nitrogen and titration analysis)
1-Amino-2,2,2-trifluoroethane hydrochloride
CAS Number: 373-88-6
MDL Number: MFCD00012875
Molecular Formula: C2H4F3N
Molecular Weight: 135.51
Purity/Analysis Method: >98.0% (N,T)
Form: Crystal
Color: White
Formula:
C₂H₅ClF₃N Melting Pt: 222…226 °C |
MDL Number:
MFCD00012875 CAS Number: 373-88-6 EINECS: 206-771-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...