α,α,α-Trifluoro-p-toluic acid ≥98.0% (by titrimetric analysis)
Supplier: TCI America
|
Warning
|
T1145-25G
T1145-5G
TCT1145-025GEA
213.3
CAD
TCT1145-025G
TCT1145-5G
α,α,α-Trifluoro-p-toluic acid ≥98.0% (by titrimetric analysis)
α,α,α-Trifluoro-p-toluic acid
CAS Number: 455-24-3
MDL Number: MFCD00002562
Molecular Formula: C8H5F3O2
Molecular Weight: 190.12
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 223
Product Categories: Synthetic Organic Chemistry, Carboxylic Acids (Multi-component Reaction), Fluorinated Building Blocks
Other Product Names / Synonyms: alpha,alpha,alpha-Trifluoro-p-toluic Acid
Formula:
C₈H₅F₃O₂ Melting Pt: 223 °C |
MDL Number:
MFCD00002562 CAS Number: 455-24-3 EINECS: 207-242-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...