2,3,4-Trimethoxybenzoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

T1116-25G T1116-5G
TCT1116-025GEA 263.86 CAD
TCT1116-025G TCT1116-5G
2,3,4-Trimethoxybenzoic acid ≥98.0% (by GC, titration analysis)
2,3,4-Trimethoxybenzoic acid

CAS Number: 573-11-5
MDL Number: MFCD00002433
Molecular Formula: C10H12O5
Molecular Weight: 212.20
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 100

Formula: C₁₀H₁₂O₅
Boiling Pt: 110…115 °C (0.04 mmHg)
Melting Pt: 100 °C
MDL Number: MFCD00002433
CAS Number: 573-11-5
EINECS: 209-350-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dd2de4d3b59e6d30d3ee3f8d5caa23bcaf639334&vid=b60191cf9cefd5dcd333ca08577b2ff1a29f992c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR