(3,4,5-Trimethoxyphenyl)acetic acid ≥98.0%

Supplier: TCI America

Synonyms: 3,4,5-Trimethoxyphenylacetic acid

T1105-5G T1105-25G
TCT1105-005GEA 86.9 CAD
TCT1105-005G TCT1105-025G
(3,4,5-Trimethoxyphenyl)acetic acid ≥98.0%
(3,4,5-Trimethoxyphenyl)acetic acid

CAS Number: 951-82-6
MDL Number: MFCD00004336
Molecular Formula: C11H14O5
Molecular Weight: 226.23
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 118

Formula: (C₆H₂(OCH₃)₃)CH₂COOH
Boiling Pt: 352 °C (1013 hPa)
Melting Pt: 118 °C
MDL Number: MFCD00004336
CAS Number: 951-82-6
EINECS: 213-456-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=71ecea83a5135c8bca4aa76d8aa1f24f455ee0de&vid=71ecea83a5135c8bca4aa76d8aa1f24f455ee0de&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR