3,4,5-Trimethoxybenzyl alcohol ≥98.0%

Supplier: TCI America

T1103-500G T1103-25G
TCT1103-500GEA 881.64 CAD
TCT1103-500G TCT1103-025G
3,4,5-Trimethoxybenzyl alcohol ≥98.0%
3,4,5-Trimethoxybenzyl alcohol

CAS Number: 3840-31-1
MDL Number: MFCD00004639
Molecular Formula: C10H14O4
Molecular Weight: 198.22
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 155
Flash Point (°C): 110
Specific Gravity (20/20): 1.20

Formula: C₁₀H₁₄O₄
Boiling Pt: 155 °C
Melting Pt: 36…38 °C
Density: 1.20 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00060356
CAS Number: 3840-31-1
EINECS: 223-331-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f295ced08085a66ca19d3faa5d14b89c3500dbfb&vid=f295ced08085a66ca19d3faa5d14b89c3500dbfb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR