1,2,3-Trimethoxybenzene ≥99.0%
Supplier: TCI America
T1102-25G
T1102-500G
TCT1102-025GEA
71.1
CAD
TCT1102-025G
TCT1102-500G
1,2,3-Trimethoxybenzene ≥99.0%
1,2,3-Trimethoxybenzene
CAS Number: 634-36-6
MDL Number: MFCD00008358
Molecular Formula: C9H12O3
Molecular Weight: 168.19
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 235
Melting point (°C): 45
Flash Point (°C): 113
Other Product Names / Synonyms: Pyrogallol Trimethyl Ether
Formula:
C₉H₁₂O₃ MW: 168.19 g/mol Boiling Pt: 235 °C Melting Pt: 45 °C Density: 1.1118 g/cm³ (45 °C) Flash Pt: 113 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008358 CAS Number: 634-36-6 EINECS: 211-207-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...