1-Iodoperfluorohexane ≥98.0%

Supplier: TCI America

T1098-25G T1098-5G
TCT1098-025GEA 126.4 CAD
TCT1098-025G TCT1098-5G
1-Iodoperfluorohexane ≥98.0%
1-Iodoperfluorohexane

CAS Number: 355-43-1
MDL Number: MFCD00001063
Molecular Formula: C6F13I
Molecular Weight: 445.95
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Very Pale Yellow
Boiling point (°C): 117
Melting point (°C): -45
Flash Point (°C): 115
Specific Gravity (20/20): 2.06


Product Categories: Synthetic Organic Chemistry, Fluorous Compounds, Materials Chemistry, Derivatizing Reagents for Solubility Improvement
Other Product Names / Synonyms: Perfluorohexyl Iodide, Tridecafluoro-1-iodohexane

Formula: C₆F₁₃I
Boiling Pt: 117 °C
Melting Pt: –45 °C
Density: 2.06 (20 °C)
Flash Pt: 115 °C
Storage Temperature: Ambient
MDL Number: MFCD00001063
CAS Number: 355-43-1
EINECS: 206-586-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8141642387847d974ff2b4b664f2b93d6e3729d3&vid=8141642387847d974ff2b4b664f2b93d6e3729d3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR