Disodium terephthalate ≥99.0%

Supplier: TCI America

Synonyms: Terephthalic acid disodium salt

T1097-25G T1097-500G T1097-100G
TCT1097-025GEA 75.57 CAD
TCT1097-025G TCT1097-500G TCT1097-100G
Disodium terephthalate ≥99.0%
Disodium terephthalate

CAS Number: 10028-70-3
MDL Number: MFCD00013137
Molecular Formula: C8H6O4
Molecular Weight: 210.10
Purity/Analysis Method: >99.0% (HPLC,T)
Form: Crystal
Color: White


Other Product Names / Synonyms: Terephthalic Acid Disodium Salt

Formula: C₈H₄Na₂O₄
MDL Number: MFCD00013137
CAS Number: 10028-70-3
EINECS: 233-075-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f11c17d2966824b8483de05ec85c2efbd9f0d423&vid=f11c17d2966824b8483de05ec85c2efbd9f0d423&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR