Methyl-3,4,5-trimethoxybenzoate ≥99.0%

Supplier: TCI America

Synonyms: 3,4,5-Trimethoxybenzoic acid methyl ester, Methyl 3,4,5-trimethoxybenzoate, Trimethylgallic Acid Methyl Ester

T1092-25G T1092-500G
TCT1092-025GEA 26.86 CAD
TCT1092-025G TCT1092-500G
Methyl-3,4,5-trimethoxybenzoate ≥99.0%
Methyl 3,4,5-rimethoxybenzoate

CAS Number: 1916-07-0
MDL Number: MFCD00008431
Molecular Formula: C11H14O5
Molecular Weight: 226.23
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 185
Melting point (°C): 82


Other Product Names / Synonyms: 3,4,5-Trimethoxybenzoic Acid Methyl Ester

Formula: C₁₁H₁₄O₅
MW: 226.23 g/mol
Boiling Pt: 185 °C
Melting Pt: 82 °C
MDL Number: MFCD00008431
CAS Number: 1916-07-0
EINECS: 217-629-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=57fe3823ead719249f8daeae798b5a5d026fd5c4&vid=57fe3823ead719249f8daeae798b5a5d026fd5c4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR