α,α,α-Trifluoro-4-tolualdehyde ≥95.0%
Supplier: TCI America
Synonyms:
4-(Trifluoromethyl)benzaldehyde, 1-Formyl-4-trifluoromethylbenzene, p-Trifluoromethylbenzaldehyde
CAS Number: 455-19-6
MDL Number: MFCD00006952
Molecular Formula: C8H5F3O
Molecular Weight: 174.12
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 64
Flash Point (°C): 79
Freezing point (°C): 1
Specific Gravity (20/20): 1.30
Product Categories: Synthetic Organic Chemistry, Aldehydes (Multi-component Reaction), Fluorinated Building Blocks
Other Product Names / Synonyms: alpha,alpha,alpha-Trifluoro-p-tolualdehyde
Formula:
C₈H₅F₃O |
MDL Number:
MFCD00006952 CAS Number: 455-19-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...