2,4,6-Tribromoaniline ≥98.0%

Supplier: TCI America

T1088-500G T1088-25G
TCT1088-500GEA 477.16 CAD
TCT1088-500G TCT1088-025G
2,4,6-Tribromoaniline ≥98.0%
2,4,6-Tribromoaniline

CAS Number: 147-82-0
MDL Number: MFCD00007634
Molecular Formula: C6H4Br3N
Molecular Weight: 329.82
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 120

Formula: C₆H₄Br₃N
MW: 329.82 g/mol
Boiling Pt: 300 °C (1013 hPa)
Melting Pt: 120 °C
Density: 2.578 g/cm³ (20 °C)
MDL Number: MFCD00007634
CAS Number: 147-82-0
EINECS: 205-700-1
UN: 2811
ADR: 6.1,II
Merck Index: 12,09740
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6dfe3735f02ed1c9c12278307941b4380d8cccfd&vid=429fd4cf9a90b9cb9b277df7a10e50c855bd35d2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR