3-Thenoic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
T1084-5G
T1084-25G
TCT1084-005GEA
115.34
CAD
TCT1084-005G
TCT1084-25G
3-Thenoic acid ≥98.0% (by GC, titration analysis)
3-Thenoic acid
CAS Number: 88-13-1
MDL Number: MFCD00005467
Molecular Formula: C5H4O2S
Molecular Weight: 128.15
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 138
Product Categories: Materials Chemistry, Conducting Polymer Research Reagents
Other Product Names / Synonyms: 3-Thenoic Acid
Formula:
C₅H₄O₂S Melting Pt: 138 °C |
MDL Number:
MFCD00005467 CAS Number: 88-13-1 EINECS: 201-802-5 Merck Index: 12,09415 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...