Trimellitic anhydride chloride ≥98.0% (by titrimetric analysis)
Supplier: TCI America
T1079-500G
T1079-25G
TCT1079-500GEA
352.34
CAD
TCT1079-500G
TCT1079-25G
Trimellitic anhydride chloride ≥98.0% (by titrimetric analysis)
Trimellitic anhydride chloride
CAS Number: 1204-28-0
MDL Number: MFCD00005924
Molecular Formula: C9H3ClO4
Molecular Weight: 210.57
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Boiling point (°C): 195
Melting point (°C): 69
Product Categories: Materials Chemistry, Monomers, Polyimide Raw Materials
Other Product Names / Synonyms: 4-Chloroformylphthalic Anhydride
Formula:
C₉H₃ClO₄ Boiling Pt: 195 °C Melting Pt: 69 °C |
MDL Number:
MFCD00005924 CAS Number: 1204-28-0 EINECS: 214-874-8 UN: 3261 ADR: 8,II |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...