4-Tolualdehyde ≥95.0%
Supplier: TCI America
|
Warning
|
T1073-25ML
T1073-500ML
TCT1073-025MLEA
40.24
CAD
TCT1073-025ML
TCT1073-500ML
4-Tolualdehyde ≥95.0%
4-Tolualdehyde
CAS Number: 104-87-0
MDL Number: MFCD00006954
Molecular Formula: C8H8O
Molecular Weight: 120.15
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 204
Flash Point (°C): 88
Specific Gravity (20/20): 1.02
Product Categories: Synthetic Organic Chemistry, Aldehydes (Multi-component Reaction)
Other Product Names / Synonyms: 4-Methylbenzaldehyde
Formula:
C₈H₈O Boiling Pt: 204 °C Melting Pt: –6 °C Density: 1.02 (20 °C) Flash Pt: 88 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006954 CAS Number: 104-87-0 EINECS: 203-246-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...