Trimethylsulfoxonium iodide ≥98.0% (by titrimetric analysis)

Supplier: TCI America

T1065-25G T1065-500G T1065-100G
TCT1065-025GEA 36.34 CAD
TCT1065-025G TCT1065-500G TCT1065-100G
Trimethylsulfoxonium iodide ≥98.0% (by titrimetric analysis)
Trimethylsulphoxonium iodide

CAS Number: 1774-47-6
MDL Number: MFCD00011899
Molecular Formula: C3H9IOS
Molecular Weight: 220.07
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White

Formula: C₃H₉IOS
Melting Pt: 165…175 °C
MDL Number: MFCD00011899
CAS Number: 1774-47-6
EINECS: 217-204-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c01a612332400bfdf41258f836f9f3797102c9e4&vid=c01a612332400bfdf41258f836f9f3797102c9e4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR