Tetraphenylcyclopentadienone ≥98.0%

Supplier: TCI America

Synonyms: 2,3,4,5-Tetraphenylcyclopenta-2,4-dienone

T1062-25G T1062-5G
TCT1062-025GEA 219.62 CAD
TCT1062-025G TCT1062-005G
Tetraphenylcyclopentadienone ≥98.0%
Tetraphenylcyclopentadienone

CAS Number: 479-33-4
MDL Number: MFCD00001407
Molecular Formula: C29H20O
Molecular Weight: 384.48
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Color: Black
Melting point (°C): 217

Formula: C₂₉H₂₀O
MW: 384.48 g/mol
Melting Pt: 217 °C
Density: 1.233 g/cm³ (20 °C)
MDL Number: MFCD00001407
CAS Number: 479-33-4
EINECS: 207-530-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=43d1593e924a2c590636c2ccdf76f8a433af3f54&vid=43d1593e924a2c590636c2ccdf76f8a433af3f54&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR