Trimethylsulfonium iodide ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

T1056-25G T1056-500G
TCT1056-025GEA 36.97 CAD
TCT1056-025G TCT1056-500G
Trimethylsulfonium iodide ≥98.0% (by titrimetric analysis)
Trimethylsulphonium iodide

CAS Number: 2181-42-2
MDL Number: MFCD00011632
Molecular Formula: C3H9IS
Molecular Weight: 204.07
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 203

Formula: C₃H₉IS
Melting Pt: 203 °C
Density: 1.922 g/cm³ (20 °C)
MDL Number: MFCD00011632
CAS Number: 2181-42-2
EINECS: 218-555-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9baa2a61e947a48d2e8be733479c1ec50fe98114&vid=9baa2a61e947a48d2e8be733479c1ec50fe98114&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR