2-Butynoic acid ethyl ester ≥98.0%

Supplier: TCI America
Warning

T1052-1ML T1052-5ML
TCT1052-001MLEA 74.26 CAD
TCT1052-001ML TCT1052-005ML
2-Butynoic acid ethyl ester ≥98.0%
2-Butynoic acid ethyl ester

CAS Number: 4341-76-8
MDL Number: MFCD00015182
Molecular Formula: C6H8O2
Molecular Weight: 112.13
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 163
Flash Point (°C): 62
Specific Gravity (20/20): 0.97


Other Product Names / Synonyms: 2-Butynoic Acid Ethyl Ester, Tetrolic Acid Ethyl Ester, Ethyl Tetrolate

Formula: C₆H₈O₂
Boiling Pt: 163 °C
Density: 0.97 (20 °C)
Flash Pt: 62 °C
MDL Number: MFCD00015182
CAS Number: 4341-76-8
EINECS: 224-395-6
UN: 3272
ADR: 3,III

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR