2,3,5,6-Tetrafluorophenol ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

T1030-25G T1030-5G
TCT1030-025GEA 254.54 CAD
TCT1030-025G TCT1030-005G
2,3,5,6-Tetrafluorophenol ≥98.0% (by titrimetric analysis)
2,3,5,6-Tetrafluorophenol

CAS Number: 769-39-1
MDL Number: MFCD00002157
Molecular Formula: C6H2F4O
Molecular Weight: 166.07
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Boiling point (°C): 146
Flash Point (°C): 79
Freezing point (°C): 35

Formula: C₆H₂F₄O
MW: 166.07 g/mol
Boiling Pt: 146 °C
Melting Pt: 30 °C
Flash Pt: 79 °C
MDL Number: MFCD00002157
CAS Number: 769-39-1
EINECS: 212-209-6
UN: 3261
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a80cf0deb9a4d62231d780cad632bc6c4b544424&vid=d55fb615668aa10686012ab03afaaca8b0db5499&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR