Tris(3-methylphenyl)phosphine ≥98.0%

Supplier: TCI America

T1025-25G T1025-5G
TCT1025-25GEA 246.32 CAD
TCT1025-25G TCT1025-005G
Tris(3-methylphenyl)phosphine ≥98.0%
Tris(3-methylphenyl)phosphine

CAS Number: 6224-63-1
MDL Number: MFCD00008532
Molecular Formula: C21H21P
Molecular Weight: 304.37
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 100


Product Categories: Synthetic Organic Chemistry, Cross-coupling Reaction using Transition Metal Catalysts, Ligands
Other Product Names / Synonyms: Tris(3-methylphenyl)phosphine

Formula: C₂₁H₂₁P
Melting Pt: 100 °C
MDL Number: MFCD00008532
CAS Number: 6224-63-1
EINECS: 228-312-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c0c5546a5f44c24a52680767097b6c256979dab2&vid=c0c5546a5f44c24a52680767097b6c256979dab2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR