Tetrahexylammonium iodide ≥98.0% (by titrimetric analysis)

Supplier: TCI America

T1010-25G T1010-5G
TCT1010-25GEA 206.66 CAD
TCT1010-25G TCT1010-005G
Tetrahexylammonium iodide ≥98.0% (by titrimetric analysis)
Tetrahexylammonium iodide

CAS Number: 2138-24-1
MDL Number: MFCD00041981
Molecular Formula: C24H52IN
Molecular Weight: 481.59
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 100

Formula: C₂₄H₅₂IN
Melting Pt: 100 °C
Density: 1.0922 g/cm³ (25 °C)
MDL Number: MFCD00041981
CAS Number: 2138-24-1
EINECS: 218-382-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aa77a6629c2b271b93af87da0eb8461431b8d572&vid=aa77a6629c2b271b93af87da0eb8461431b8d572&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR