2,4,5-Triphenylimidazole ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: Lophine, 2,4,5-Triphenyl-1H-imidazole

T0999-25G T0999-5G
TCT0999-025GEA 112.18 CAD
TCT0999-025G TCT0999-005G
2,4,5-Triphenylimidazole ≥98.0% (by HPLC, titration analysis)
2,4,5-Triphenylimidazole

CAS Number: 484-47-9
MDL Number: MFCD00005187
Molecular Formula: C21H16N2
Molecular Weight: 296.37
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: White
Melting point (°C): 275
Lambda max.: 300 nm (EtOH)


Other Product Names / Synonyms: Lophine

Formula: C₂₁H₁₆N₂
MW: 296.37 g/mol
Boiling Pt: 509 °C (1013 hPa)
Melting Pt: 275 °C
MDL Number: MFCD00005187
CAS Number: 484-47-9
EINECS: 207-606-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fa1caa998c1c82597a66a909b90ca77dba7f9bc2&vid=fa1caa998c1c82597a66a909b90ca77dba7f9bc2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR